Learn More
Ethos Biosciences Acid Alcohol 0.5%, 1gal, Ethos
Supplier:  Ethos Biosciences 7560G
Encompass_Preferred
Acid Alcohol 0.5% is a ready-to-use differentiating solution formulated for controlled decolorization in routine staining procedures. Typically prepared as a dilute hydrochloric acid solution in alcohol, it is used to selectively remove excess dye from tissue sections or cytology smears, enhancing contrast and definition of stained structures. Commonly applied in hematoxylin-based protocols and other staining methods requiring precise differentiation, this solution supports consistent, reproducible results. Suitable for use in clinical, research, and educational laboratory environments.
Catalog No. 50-005-74267