Learn More
Thomas Scientific Reagent Reservoir, 25 ml, Sterile, Individually Wrapped, 100/CS
SDP
Supplier:  Thomas Scientific 1228K18
Encompass
“V” bottom design allows for maximum recovery of reagentsGraduated with convenient pouring spouts at all four cornersExtra wide base for increased stabilityUnique “Nesting” feature makes separation of bulk packed reservoirs easy
Catalog No. NC2753394