Learn More
Lonza Walkersville LAL Reagent Reservoirs
Supplier:  Lonza Walkersville 00190035
Encompass
LAL reagent reservoirs can be used with multi channel pipettors when adding LAL reagents to a 96-well plate. The LAL reagent reservoirs are provided in a zip closure bag enabling you to conveniently store unused reservoirs for later use. The LAL reagent reservoirs are tested for endotoxin and must meet a specification of <0.005 EU/ml.Benefits:Certified to contain less than 0.005 EU/mlContent:10/pack
Catalog No. 50-113-1960