Learn More
Hach Company Chloride Reagent Set, Silver Nitrate Digital Titrator, Full Range
Supplier:  Hach Company 2288000
Encompass
For determination of chloride by Silver Nitrate Digital Titrator Titration. Hach Method 8207. Ranges: 10-100 and 100-10,000 mg/L as CI- . Set contains: Chloride 2 Indicator Powder Pillow packs and two Silver Nitrate Titration Cartridges (0.2256 N and 1.128 N). Approximately 40 to 100 tests per range.
Catalog No. 50-199-5741