Learn More
Hach Company Copper Reagent Set, Free and Total
Supplier:  Hach Company 2439200
Encompass
For determination of free and complexed (chelated) Copper by the Binchoninate method. Range: up to 5.00 mg/L. Set includes: Free Copper Reagent Powder Pillows and Hydrosulfite Power Pillows for 100 tests.
Catalog No. 50-199-5998