Learn More
Hach Company Oxygen Scavenger Reagent Set
Supplier:  Hach Company 2446600
Encompass
For determination of oxygen scavengers by the iron reduction method. Hach method 8140 determines carbohydrazide, diethylhydroxylamine (DEHA), hydroquinone, iso-ascorbic acid (erythorbic acid) and methylethylketoxime. Set contains DEHA 1 Reagent Powder Pillows and DEHA 2 Solution for 100 tests
Catalog No. 50-199-6013