Learn More
Hach Company Hardness (Total) Reagent Set, Buret, LR
Supplier:  Hach Company 2447600
Encompass
Reagent set for the determination of Total Hardness by the ManVer 2 Buret Titration Method (Hach method 8226). EPA Accepted method. Range: 0-2,500 mg/L as CaCO3. Contains: Buffer Solution (Hardness 1), ManVer 2 Hardness Indicator Powder Pillows and 0.0200 N EDTA (TitraVer) Titrant for up to approximately 100 tests.
Catalog No. 50-199-6022