Learn More
Hach Company Molybdenum Reagent Set, HR, 25 mL
Supplier:  Hach Company 2243400
Encompass
For determination of Molybdenum by the Mercaptoacetic Acid Method (Hach method 8036). Range: Up to 35.0 mg/L as Mo6+. Sample Size: 25 mL. Set Contains: MolyVer 1, 2 and 3 Molybdenum Reagent Powder Pillows for 100 tests.
Catalog No. 50-199-5620