Learn More
Hach Company Nitrogen-Ammonia Set, Salicylate, 25 mL
Supplier:  Hach Company 2243700
Encompass
For determination of Ammonia Nitrogen by the Salicylate Method. Hach Method 8155. Range: 0.01 - 0-0.50 mg/L as NH3-N. Sample Size: 25 mL. Set contains Ammonia Salicylate Powder Pillows and Ammonia Cyanurate Powder Pillows for 100 tests.
Catalog No. 50-199-5622