Learn More
Hach Company Tannin-Lignin Reagent Set
Supplier:  Hach Company 2244600
Encompass
For determination of Tannin and Lignin by the Tyrosine Method. Hach Method 8193. Range: 0.1 - 9.0 mg/L. Sample Size: 25 mL. Set Includes: Sodium Carbonate Solution and TanniVer™ 3 Tannin-Lignin Reagent for up to 100 tests.
Catalog No. 50-199-5630