Learn More
Hach Company Hardness (Total) Reagent Set, Digital Titrator, Full Range
Supplier:  Hach Company 2272000
Encompass
For Total Hardness determination by EDTA Digital Titrator titration with Calmagite indicator. Range: 10-160 and 100-4000 mg/L as CaCO3. Set contains ManVer 2 Indicator, Hardness 1 Buffer Solution and one cartridge each of 0.0800 M and 0.800 M EDTA.
Catalog No. 50-199-5691