Learn More
Hach Company Hardness (ULR) Reagent Set, Bulk Solution
Supplier:  Hach Company 2603101
Encompass
Reagent set ultra low range hardness determination by the Chlorophosphonazo method. Hach Method 8374. Range: 4 - 1000 ug/L Ca & Mg as CaCO3. Chlorophosphonazo reagent solution is packaged in a 500-mL bottle that is ideal for use with Hach's Pour-Thru Cell. Approximately 250 tests
Catalog No. 50-199-6313