Learn More
Medchemexpress LLC HY-B0360 500mg Medchemexpress, Rebamipide CAS:90098-04-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB0360500MG
Encompass_Preferred
Medchemexpress, HY-B0360 500mg Rebamipide CAS:90098-04-7 Rebamipide (OPC12759) is a mucoprotective agent. Rebamipide induces COX-2 expression, increases PGE2 levels, and enhances gastric mucosal defense in a COX-2-dependent manner. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2489