Learn More
Medchemexpress LLC Rebamipide impurity 13 | 4876-17-9 | MFCD22194962 | >98.0% | 193.63 g/mol | C10H8ClNO | 5 MG

Supplier: Medchemexpress LLC HYW2509955MG
Rebamipide impurity 13 is a small-molecule impurity derived from the gastroprotective agent rebamipide, supplied for research and drug-intermediate uses. It is characterized by a chloroquinolinone core and is used in analytical impurity profiling and as a synthetic intermediate in medicinal chemistry research.
- CAS number 4876-17-9.
- Molecular formula C10H8ClNO; molecular weight 193.63 g/mol.
- Commonly reported ≥98.0% purity by distributor specifications.
- Intended use drug intermediate and impurity reference in analytical studies.
- Storage recommendations: powder at -20°C or 4°C; solutions at -80°C or -20°C.
- Available in small research sizes such as 5 mg and 10 mg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.