Learn More
Medchemexpress LLC Rebastinib | 1020172-07-9 | 99.8% | 100 MG
SDP
Supplier:  Medchemexpress LLC HY13024100MG
Encompass_Preferred
Rebastinib (DCC-2036) is an orally active, non-ATP-competitive inhibitor targeting Bcr-Abl, including resistant mutants. It effectively inhibits Abl1WT and Abl1T315I, as well as SRC, KDR, FLT3, and Tie-2, with low activity against c-Kit. This compound demonstrates potent antiproliferative effects in various leukemia cell lines and significantly prolongs survival in animal models of leukemia.
  • Potently inhibits ABL1 native and ABL1 T315I.
  • Inhibits SRC family kinases and PDGFRα/β.
  • Selectively spares c-KIT.
  • Effectively inhibits proliferation of native BCR-ABL1 and Ph+ K562 cell lines.
  • Inhibits proliferation of common TKI-resistant BCR-ABL1 mutants.
  • Reduces leukemia cell burden in treated mice.
Catalog No. 50-003-40053