Learn More
Selleck Chemical LLC Rebastinib10mg
SDP
Supplier:  Selleck Chemical LLC S263410MG
Encompass_Preferred
Rebastinib (DCC-2036) is a conformational control Bcr-Abl inhibitor for Abl1(WT) and Abl1(T315I) with IC50 of 0.8 nM and 4 nM, also inhibits SRC, LYN, FGR, HCK, KDR, FLT3, and Tie-2, and low activity to seen towards c-Kit. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3092