Learn More
Cayman Chemical RebaudIosIde A 50g
SDP
Supplier:  Cayman Chemical 1189450 G
Encompass_Preferred
A natural non-caloric sweetener; increases GLP-1 secretion in a 2-D mouse intestine model; mice prefer rebaudioside A water to unsweetened water
Catalog No. 50-295-3058