Learn More
Medchemexpress LLC Rebaudioside M | 1220616-44-3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN683310MM/1ML
Encompass_Preferred
Rebaudioside M is a non-calorie sweetener isolated from Stevia rebaudiana. This glycoside of the ent-kaurene diterpenoid aglycone is over 350 times sweeter than sucrose and is suitable for use in food and beverages. It has also been observed to provide antidiabetic, antihypertensive, anti-inflammatory, antioxidant, anticaries, and anticancer benefits.
  • Protects beta-cell activity and enhances insulin secretion
  • Inhibits the PI3K/Akt signaling pathway
  • Downregulates the Wnt/catenin signaling pathway in colon cancer cells
  • Lowers fasting blood glucose and enhances glucose tolerance
  • Decreases oxidative stress and pro-inflammatory cytokines
  • Reduces blood pressure and increases endothelial function
Catalog No. 50-004-17905