Learn More
Sigma Aldrich Fine Chemicals Biosciences Rebaudioside M 95.0 HPLC50MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 7463250MG
Encompass
Rebaudioside M is a diterpene glycoside isolated from Stevia rebaudiana leaves. Rebaudioside M is a zero-calorie sweetener and has a clean sweet taste with slight bitter or licorine aftertaste.
Catalog No. 50-289-8066