Learn More
Chromadex Rebaudioside M(P) | 1220616-44-3 | MFCD31916341 | 25mg
SDP
Supplier:  Chromadex ASB00018234025
Encompass_Preferred
Rebaudioside M(P) | Putity: 96.3 | MW: 1291.29 | 1220616-44-3 | MFCD31916341 | 25mg
Catalog No. NC1613807