Learn More
Medchemexpress LLC Rebaudioside M (Standard) | 1220616-44-3 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYN6833R10MG
Encompass_Preferred
Rebaudioside M (Standard) is the analytical standard of Rebaudioside M, a non-calorie sweetener isolated from *Stevia rebaudiana* that is more than 350 times sweeter than sucrose. Intended for research and analytical applications, this compound is of analytical standard grade.
  • Used as a reference standard assay.
  • Commonly utilized in qualitative, quantitative, and methodological research experiments.
  • Suitable for HPLC, GC, and MS applications.
  • Appears as a white to off-white solid.
  • For research use only.
Catalog No. 50-004-19487