Learn More
Medchemexpress LLC Reboxetine mesylate | 98769-84-7 | MFCD02184016 | ≥99.0% | C20H27NO6S | 10MG

Supplier: Medchemexpress LLC HY14560C10MG
Reboxetine mesylate is the methanesulfonate salt of reboxetine, a selective noradrenaline reuptake inhibitor used in research to study norepinephrine transporter pharmacology and antidepressant mechanisms.
- Selective noradrenaline reuptake inhibitor activity.
- Useful for research on antidepressant pharmacology and noradrenergic signaling.
- Reported purity ≥99.0% (supplier listings).
- Chemical formula C20H27NO6S; molecular weight 409.50.
- Recommended storage: 4°C; in solvent -80°C (6 months) or -20°C (1 month).
- Suitable as a research reagent for in vitro and in vivo studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.