Learn More
Biotang Inc Batroxobin Recombinant, Produced in yeast, glycosilated polypeptide chain containing 213 aa and having a 28-33 kDa.Synonyms: EC 3.4.21.74, BX, Bothrops atrox serine proteinase, Venombin-A, Defibrase, Reptilase, Batroxobin
Supplier:  Biotang Inc BTPRO607
Encompass_Preferred
Batroxobin Recombinant, Produced in yeast, glycosilated polypeptide chain containing 213 aa and having a 28-33 kDa.Synonyms: EC 3.4.21.74, BX, Bothrops atrox serine proteinase, Venombin-A, Defibrase, Reptilase, Batroxobin
Catalog No. 50-148-927