Learn More
Biotang Inc Elcatonin, Elcatonin Synthetic is a single, non-glycosylated polypeptide chain containing 31 amino acids, having a molecular mass of 3363.2 Dalton and a Molecular formula of C148H244N42O47.
Supplier:  Biotang Inc BTHOR302
Encompass_Preferred
Elcatonin, Elcatonin Synthetic is a single, non-glycosylated polypeptide chain containing 31 amino acids, having a molecular mass of 3363.2 Dalton and a Molecular formula of C148H244N42O47.
Catalog No. 50-149-016