Learn More
Abcam Recombinant Human SPAM1 protein, 2UG
Supplier:  Abcam AB1322582UG
Encompass_Preferred
Recombinant Human SPAM1 protein is a Human Full Length protein, in the 1 to 509 aa range, expressed in Wheat germ and suitable for SDS-PAGE, ELISA, WB.
Protein/ Peptide/ Enzyme Synonyms: HYAL3, PH20, SPAM1, Hyaluronidase PH-20, Hyal-PH20, Hyaluronoglucosaminidase PH-20, Sperm adhesion molecule 1, Sperm surface protein PH-20
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1966615