Learn More
Biotang Inc Goserelin, Greater than 98.0% Goserelin contains 10 amino acids Glu1-His2-Trp3-Ser4-Tyr5-D-Ser(tBu)6-Leu7-Arg8-Pro9-AzGly10-NH2 and having a molecular weight of 1269.43 Dalton
Supplier:  Biotang Inc BTHOR256
Encompass_Preferred
Goserelin, Greater than 98.0% Goserelin contains 10 amino acids Glu1-His2-Trp3-Ser4-Tyr5-D-Ser(tBu)6-Leu7-Arg8-Pro9-AzGly10-NH2 and having a molecular weight of 1269.43 Dalton
Catalog No. 50-149-701