Learn More
Biotang Inc HIV-1 p24 gag Recombinant, HIV-1 p24 gag, containing the HIV-1 p24 gag immunodominant regions, 77-436 amino acids.The HIV-1 p24 gag is fused to beta-galactosidase (114 kDa).
Supplier:  Biotang Inc BTHIV123
Encompass_Preferred
HIV-1 p24 gag Recombinant, HIV-1 p24 gag, containing the HIV-1 p24 gag immunodominant regions, 77-436 amino acids.The HIV-1 p24 gag is fused to beta-galactosidase (114 kDa).
Catalog No. 50-151-681