Learn More
Biotang Inc Klotho Human Recombinant, KLOTHO Human,The Klotho Human Recombinant is produced in CHO cells and fused with a C-terminal 6xHis Tag. The Klotho His Tagged Fusion Protein is 59.5kDa protein containing a total of 522 aa
Supplier:  Biotang Inc BTENZ369
Encompass_Preferred
Klotho Human Recombinant, KLOTHO Human,The Klotho Human Recombinant is produced in CHO cells and fused with a C-terminal 6xHis Tag. The Klotho His Tagged Fusion Protein is 59.5kDa protein containing a total of 522 aa
Catalog No. 50-149-953