Learn More
Biotang Inc Myostatin Human Recombinant, His Tag, Total 152AA. M.W. 16.7kDa .N-terminal His-tag and spacer (43AA ). The AA sequence of the human myostatin part of the fusion protein is corresponding to the UniProtKB/Swiss-Prot entry O14793.
Supplier:  Biotang Inc BTCYT445
Encompass_Preferred
Myostatin Human Recombinant, His Tag, Total 152AA. M.W. 16.7kDa .N-terminal His-tag and spacer (43AA ). The AA sequence of the human myostatin part of the fusion protein is corresponding to the UniProtKB/Swiss-Prot entry O14793.
Catalog No. 50-150-104