Learn More
Biotang Inc Placental Lactogen Human Recombinant, CSH1, Choriomammotropin, Lactogen, CSH2, PL, CSA, CSMT. containing 199 amino acids and an additional Ala at the N-terminus having a molecular mass of approximately 22.4 kDa.
Supplier:  Biotang Inc BTCYT656
Encompass_Preferred
Placental Lactogen Human Recombinant, CSH1, Choriomammotropin, Lactogen, CSH2, PL, CSA, CSMT. containing 199 amino acids and an additional Ala at the N-terminus having a molecular mass of approximately 22.4 kDa.
Catalog No. 50-150-250