Learn More
Biotang Inc Procalcitonin Human Recombinant (PCT), His Tag, containing 114 amino acids fragment (3-116) having a molecular mass of 17.13 kDa and an amino-terminal hexahistidine tag.
Supplier:  Biotang Inc BTHOR295
Encompass_Preferred
Procalcitonin Human Recombinant (PCT), His Tag, containing 114 amino acids fragment (3-116) having a molecular mass of 17.13 kDa and an amino-terminal hexahistidine tag.
Catalog No. 50-150-278