Learn More
Biotang Inc Trypsin Human Recombinant, 10,000 BAEE units/mg powder. Greater than 90% as determined by SDS-PAGE. free from protease inhibitors such as PMSF and EDTA.
Supplier:  Biotang Inc BTPRO770
Encompass_Preferred
Trypsin Human Recombinant, 10,000 BAEE units/mg powder. Greater than 90% as determined by SDS-PAGE. free from protease inhibitors such as PMSF and EDTA.
Catalog No. 50-150-656