Learn More
Biotang Inc Lipase-A Recombinant, is expressed in E.Coli having a Mw of 65 kDa, Greater than 90% as determined by SDS-PAGE. Biological Activity 580 units/mg powder.
Supplier:  Biotang Inc BTENZ473
Encompass_Preferred
Lipase-A Recombinant, is expressed in E.Coli having a Mw of 65 kDa, Greater than 90% as determined by SDS-PAGE. Biological Activity 580 units/mg powder.
Catalog No. 50-150-782