Learn More
Biotang Inc Noggin Mouse Recombinant, Synonyms Noggin, SYM1, SYNS1, NOG.is a non-glycosylated, non-disulfide-linked homodimer consisting of two 206 amino acid polypeptide chains, having a total molecular mass of approximately 46.2 kDa
Supplier:  Biotang Inc BTCYT600
Encompass_Preferred
Noggin Mouse Recombinant, Synonyms Noggin, SYM1, SYNS1, NOG.is a non-glycosylated, non-disulfide-linked homodimer consisting of two 206 amino acid polypeptide chains, having a total molecular mass of approximately 46.2 kDa
Catalog No. 50-150-899