Learn More
Biotang Inc Protein G Recombinant, The Protein G contains amino acids 190-384 having a molecular mass of 21.6 kDa. The Protein-G migrates on SDS-PAGE around 32 kDa.
Supplier:  Biotang Inc BTPRO402
Encompass_Preferred
Protein G Recombinant, The Protein G contains amino acids 190-384 having a molecular mass of 21.6 kDa. The Protein-G migrates on SDS-PAGE around 32 kDa.
Catalog No. 50-150-985
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.