Learn More
Biotang Inc Tropic 1808 Rat Recombinant, Tpc1808 Rat, produced in E.Coli is a single, non-glycosylated polypeptide chain containing 285 amino acids and having a molecular mass of 29.1 kDa.fused to N-terminal His-Tag
Supplier:  Biotang Inc BTPRO587
Encompass_Preferred
Tropic 1808 Rat Recombinant, Tpc1808 Rat, produced in E.Coli is a single, non-glycosylated polypeptide chain containing 285 amino acids and having a molecular mass of 29.1 kDa.fused to N-terminal His-Tag
Catalog No. 50-151-048