Learn More
New England Biolabs, Inc. RecA – 200 µg
SDP
Supplier:  New England Biolabs, Inc. M0249S
Encompass
E. coli RecA is necessary for genetic recombination, reactions involving DNA repair and UV-induced mutagenesis. RecA promotes the autodigestion of the lexA repressor, umuD protein and lambda repressor. Cleavage of LexA derepresses more than 20 genes. In vitro studies indicate that in the presence of ATP, RecA promotes the strand exchange of single-strand DNA fragments with homologous duplex DNA. The reaction has three distinct steps: (i) RecA polymerizes on the single-strand DNA, (ii) the nucleoprotein filament binds the duplex DNA and searches for a homologous region, (iii) the strands are exchanged.
Catalog No. NC0117510