Learn More
Fritsch Milling And Sizing Inc Receiver Unit - Board for Inserting In Planetary Mills Premium Line
SDP
Supplier:  Fritsch Milling And Sizing Inc 81.0013.00
Encompass
Turn your Planetary Mill PULVERISETTE 5 premium line and Planetary Micro Mill PULVERISETTE 7 premium line into an analytical measuring system by using the EASY GTM system (Gas Pressure and Temperature Measurement System) with special lid and transmitter as well as a receiver board and the included MillControl software.
Your advantage: easy and safe monitoring and analysis of thermal effects, physical and chemical reactions or increases/decreases in pressure through continuous measurement of gas pressure and temperature directly in the grinding bowl. The mill is automatically controlled to ensure that the set parameters are not exceeded.
Especially practical: The receiver unit needs only to be inserted and adjusted just once – the software MillControl via USB only installed once - regardless of the number of EASY GTM-bowls used.
Catalog No. 50-153-4703