Learn More
Chemglass Life Sciences Chemglass Receiving Flask, 3 L - For rotary evaporator (CG-1334-X54)
SDP
Supplier:  Chemglass Life Sciences CG1334X54
Encompass_Preferred
Receiving flask designed for rotary evaporators. Provides a 3-liter glass collection vessel to capture condensate during evaporation processes.
  • Capacity: 3 L.
  • Designed to collect condensate from a rotary evaporator.
  • Glass construction suitable for laboratory use.
  • Used as the receiving vessel in rotary evaporation setups.
Catalog No. 50-273-3595