Learn More
Innovative Research Inc Human Receptor Associated (RAP) Purified Low Endotoxin 500ug
Supplier:  Innovative Research Inc IHURAPLX500UG
Encompass_Preferred
Human Receptor Associated (RAP) Purified Low Endotoxin from Innovative Research has been recombinantly produced in E. coli and purified using a combination of conventional and affinity chromatography then endotoxin depleted and sterile filtered. This is a frozen liquid buffered in 0.0067M Phosphate; 0.15M NaCl; pH 7.4, with a concentration of 2.5 mg/ml and a purity of >95%.
Catalog No. 50-203-6491