Learn More
Selleck Chemical LLC Recilisib E2947-25MG
SDP
Supplier:  Selleck Chemical LLC E294725MG
Encompass_Preferred
Recilisib (ON01210 EX-RAD) is a radioprotectant that activates the activity of AKT and PI3K in cells It has been studied as prophylactic (use prior to radiation exposure) and therapeutic (after exposure to radiation) drug
Catalog No. 50-000-39694