Learn More
Being Scientific Recirculating Chiller 0.3 cuft (8L) 120V, 900W Cooling capacity
Supplier:  Being Scientific BRC15105U
Encompass_Preferred
The BEING BRC Series cooling/heating recirculating chillers provide continuously reliable and stable laboratory temperature control for a wide range of applications. The recirculating chillers have a bright, easy-to-understand LCD intelligent controller, a corrosion-resistant stainless steel chamber, and many more features. Available in 3 L, 10 L, and 20 L tank capacity with a temperature range from -20oC to 20oC.
Catalog No. 50-002-17549