Learn More
Enterprise Technology Solutions Reckitt Benckiser Prof Oven/Grill Cleaner, 64oz., Clear
SDP
Supplier:  Enterprise Technology Solutions RAC80689
Encompass_Preferred
This nonflammable, thick formula clings to vertical surfaces and quickly removes baked-on grease. Ready-to-use Oven and Grill Cleaner is effective on hot grills up to 170 degrees Fahrenheit - no need to shut down grill. Use on ovens, grills, griddles, broilers and vent hoods. Professional cleaner comes in a plastic bottle with a screw-top lid and easy-grip handle.
Catalog No. 50-007-89826