Learn More
Grainger Reclosable Fastener 1 x 2 In Clear PK100
Supplier:  Grainger 16U703
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Strip is a reclosable fastener that creates a strong, secure hold. It is made of continuous molded plastic stems with mushroom shaped tops that interlock when pressed together. This fastener strip has an adhesive backing for attachment to surfaces. It can be cut to desired lengths. The Dual Lock fastener is available in various widths, colors, and attachment styles. It bonds on contact and can be opened and closed repeatedly while maintaining a strong hold. This product is ideal for attaching, mounting or fastening items where a secure hold is needed.
Catalog No. 50-276-6504