Learn More
Grainger Reclosable Fastener 1 x 3 In Clear PK100
Supplier:  Grainger 16U704
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Strip is a heavy duty reclosable fastener that creates a strong, secure bond. It features a mushroom-shaped stem design that interlocks with another strip to form a connection. This fastener strip has an adhesive backing for attachment to surfaces. It can withstand up to 7 lbs/in2 shear and 40 lbs/in2 peel strength. The strip comes in black color and measures 0.75 in. wide x 10 ft. long. It is ideal for attaching two rigid surfaces together when a strong, reclosable bond is needed. This product is manufactured in the USA by 3M.
Catalog No. 50-276-6619