Learn More
Grainger Reclosable Fastener 1x1 In PK100
Supplier:  Grainger 16U708
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Square is a reclosable fastener with a strong hold, featuring a mushroom-shaped stem that interlocks for a secure bond. Made from nylon, it comes in black, white, or clear and operates from -30°F to 160°F, performing well in harsh environments. It can be cut-to-size and bonds on contact for quick fastening. Versatile for industrial, commercial, and consumer use, it is available in various shapes and sizes.
Catalog No. 50-276-6715