Learn More
Grainger Reclosable Fastener Dual Lock 30 ft.
Supplier:  Grainger 16U578
Encompass_Preferred
The 3M Dual Lock Hook and Loop Fastener Roll is a reclosable fastener that creates a strong, secure hold. It has a mushroom-shaped stem that engages with a continuous backing to provide high strength and shear resistance. This versatile fastener can bond two rigid surfaces together or act as a mechanical fastener with one adhesive side. It is available in a 1" x 25' roll in black, and has a temperature resistance range of -40°F to 220°F. The Dual Lock fastener is ideal for use in industrial, electronic, automotive, and interior design applications.
Catalog No. 50-276-7662