Learn More
Grainger Reclosable Fastener Hook 30 ft.
Supplier:  Grainger 16U576
Encompass_Preferred
The 3M Reclosable Fastener Rubber is a self-adhesive rubber fastener that can be opened and closed repeatedly for easy access. It has a peel and stick acrylic adhesive that adheres to most surfaces. The fastener measures 2 inches wide x 36 yards long and comes in black color. It can withstand temperatures from -20°F to 160°F. The rubber is flexible and waterproof. This reclosable fastener is ideal for use on tarps, covers, awnings, boats, RVs, vehicles, packaging, and other applications where repeated fastening and unfastening is needed. It provides a quick and easy way to securely close flaps and openings while still allowing convenient access.
Catalog No. 50-276-5802