Learn More
Grainger Reclosable Fastener Loop 30 ft.
Supplier:  Grainger 16U581
Encompass_Preferred
The 3M Dual Lock Reclosable Fastener is an acrylic-based fastener that creates a strong, secure bond. It features mushroom-shaped stems that interlock when pressed together, providing a connection that resists shear and peel forces. This versatile fastener can replace screws, rivets, and other permanent fasteners and allows for repeated fastening and unfastening. It is available in a variety of colors, sizes, and attachment types including tapes, strips, and dots. Key features include high strength, vibration resistance, and the ability to be opened and closed repeatedly without losing effectiveness. This fastener has many industrial, commercial, and consumer applications.
Catalog No. 50-276-5050