Learn More
Grainger Reclosable Fastener Loop 30 ft.
Supplier:  Grainger 16U571
Encompass_Preferred
The 3M Reclosable Fastener Loop 16U571 is a hook and loop fastener tape made of nylon that can be cut to size. It has an adhesive backing for attachment to surfaces. This product has a width of 1" and comes in a 25 yard roll. It can withstand temperatures up to 200°F. The loop side adheres to hook tape for a quick fastening and releasing capability. This product is designed for attaching signs, wire and cable ties, trim, and more. It can be used for both indoor and outdoor applications.
Catalog No. 50-276-7706